Assay Details
Gene Symbol: Galnt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167981.1 | NP_001161453.1 | 9 - 10 | 1682 | 92 | |
NM_144731.4 | NP_653332.3 | 9 - 10 | 1682 | 92 | ||
GenBank mRNA | AF349573.1 | - | 9 - 10 | 1606 | 92 | |
AK030683.1 | - | 9 - 10 | 1630 | 92 | ||
AK033427.1 | - | 9 - 10 | 1631 | 92 | ||
AK036523.1 | - | 9 - 10 | 1631 | 92 | ||
AK151424.1 | - | 8 - 9 | 1273 | 92 | ||
AK151753.1 | - | 8 - 9 | 1273 | 92 | ||
AK152205.1 | - | 5 - 6 | 685 | 92 | ||
AK153143.1 | - | 8 - 9 | 1273 | 92 | ||
BC007484.1 | - | 5 - 6 | 720 | 92 | ||
BC049907.1 | - | 5 - 6 | 729 | 92 | ||
BC052461.1 | - | 9 - 10 | 1658 | 6834581 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
