Assay Details
| Gene Symbol: Apoa1bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144897.3 | NP_659146.1 | 4 - 5 | 501 | 76 | |
| GenBank mRNA | AJ344092.1 | - | 4 - 5 | 501 | 76 | |
| AK131615.1 | - | 4 - 5 | 712 | 76 | ||
| AK159846.1 | - | 4 - 5 | 503 | 76 | ||
| AK178551.1 | - | 2 - 3 | 164 | 76 | ||
| AK181667.1 | - | 2 - 3 | 164 | 76 | ||
| AY566271.1 | - | 4 - 5 | 501 | 76 | ||
| BC058362.1 | - | 4 - 5 | 499 | 6819459 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Apoa1bp
Genomic Map