Assay Details
Gene Symbol: Med17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144933.1 | NP_659182.1 | 9 - 10 | 1529 | 118 | |
GenBank mRNA | AK033151.1 | - | 9 - 10 | 1535 | 118 | |
AK038451.1 | - | 9 - 10 | 1569 | 118 | ||
AK049112.1 | - | 9 - 10 | 1569 | 118 | ||
AK142033.1 | - | 9 - 10 | 1568 | 118 | ||
AK151377.1 | - | 9 - 10 | 1569 | 118 | ||
BC004062.1 | - | 8 - 9 | 1206 | 118 | ||
BC020185.1 | - | 9 - 10 | 1529 | 4505959 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
