Assay Details
Gene Symbol: Csad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144942.4 | NP_659191.1 | 3 - 4 | 425 | 67 | |
GenBank mRNA | AK005015.1 | - | 3 - 4 | 426 | 67 | |
AY033912.1 | - | 3 - 4 | 342 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
