Assay Details
Gene Symbol: Pomt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145145.1 | NP_660127.1 | 11 - 12 | 1175 | 95 | |
GenBank mRNA | AK053889.1 | - | 10 - 11 | 1695 | 95 | |
AK134770.1 | - | 11 - 12 | 1219 | 95 | ||
AK153984.1 | - | 11 - 12 | 1274 | 95 | ||
AY007238.1 | - | 11 - 12 | 1215 | 95 | ||
AY494857.1 | - | 11 - 12 | 1235 | 95 | ||
BC027325.1 | - | 11 - 12 | 1175 | 3981366 | 95 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK053889.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
