Assay Details
Gene Symbol: Tspan10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145363.2 | NP_663338.2 | 1 - 2 | 186 | 85 | |
GenBank mRNA | AK084214.1 | - | 1 - 2 | 133 | 85 | |
AK148037.1 | - | 1 - 2 | 129 | 85 | ||
AK148326.1 | - | 1 - 2 | 122 | 85 | ||
AK148383.1 | - | 1 - 2 | 122 | 85 | ||
AK211049.1 | - | 1 - 2 | 119 | 85 | ||
BC019991.1 | - | 1 - 2 | 124 | 4189407 | 85 | |
CK625019.1 | - | 1 - 2 | 186 | 85 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK148037.1,AK148326.1,AK148383.1,BC019991.1,NM_145363.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057706 | AK208499.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057706 | AK214141.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057706 | BC019991.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057706 | AK148326.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057706 | AK148155.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195874 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322400 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P148223 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173395 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057706 | AK208499.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057706 | AK214141.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057706 | BC019991.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057706 | AK148326.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057706 | AK148155.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184572 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184579 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184581 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184584 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184568 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184578 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184573 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184565 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184576 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184570 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184582 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28184567 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173395 | - | - |
Whole Mouse Genome G4122A | A_51_P176811 | AK208499.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P176811 | AK214141.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P176811 | BC019991.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P176811 | AK148326.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P176811 | AK148155.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115769 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173395 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09500328 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184581 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184517 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184613 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184658 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184534 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184561 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184536 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184539 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184520 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184544 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184586 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28184656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184579 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184590 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09427398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184592 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184598 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184561 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09500357 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09725932 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184576 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184588 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09500331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09500356 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184584 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184594 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28184578 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10383354 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383356 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383357 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426588_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10383353 | - | - |
Mouse Genome 430 2.0 Array | 1426588_at | - | - |
Mouse Genome 430 2.0 Array | 1426588_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208634_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 208634_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100019454.mm.1 | NM_145363.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019454.mm.1 | NM_145363.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019454.mm.1 | TC1100002026.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100019459.mm.1 | NM_145363.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019459.mm.1 | NM_145363.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019459.mm.1 | TC1100002026.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100019458.mm.1 | NM_145363.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019458.mm.1 | NM_145363.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019458.mm.1 | TC1100002026.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100019456.mm.1 | NM_145363.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019456.mm.1 | NM_145363.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100019456.mm.1 | TC1100002026.mm.1 | MTA TC |
Mouse Expression Set 430 | 1426588_at | - | - |
Mouse Expression Set 430 | 1426588_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1426588_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1426588_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426588_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426588_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 607511 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 7050358 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0007050358 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2661035 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2661035 | - | - |
mouse-6 v1.1 (Illumina) | scl40630.4.546_13-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 7050358 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl40630.4.546_13-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 7050358 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2661035 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2661035 | - | - |
