Assay Details
| Gene Symbol: Nsun5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145414.2 | NP_663389.2 | 2 - 3 | 245 | 67 | |
| GenBank mRNA | AF412029.1 | - | 2 - 3 | 238 | 67 | |
| AK079400.1 | - | 2 - 3 | 246 | 67 | ||
| AK149521.1 | - | 2 - 3 | 227 | 67 | ||
| AK156651.1 | - | 2 - 3 | 240 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsun5
Genomic Map