Assay Details
Gene Symbol: Kri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145416.3 | NP_663391.2 | 13 - 14 | 1290 | 60 | |
GenBank mRNA | BC021438.1 | - | 13 - 14 | 1280 | 5039543 | 60 |
CO426749.1 | - | 5 - 6 | 522 | 60 | ||
CX562209.1 | - | 3 - 4 | 317 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
