Assay Details
Gene Symbol: Kri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145416.3 | NP_663391.2 | 16 - 17 | 1641 | 69 | |
GenBank mRNA | BC021438.1 | - | 16 - 17 | 1631 | 5039543 | 69 |
CX562209.1 | - | 6 - 7 | 668 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
