Assay Details
Gene Symbol: Kri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145416.3 | NP_663391.2 | 17 - 18 | 1709 | 59 | |
GenBank mRNA | BC021438.1 | - | 17 - 18 | 1699 | 5039543 | 59 |
BI989088.1 | - | 2 - 3 | 210 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
