Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100008324.mm.1 |
NM_001271358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008324.mm.1 |
NM_001271359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008324.mm.1 |
NM_001271360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008324.mm.1 |
NM_145429.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008324.mm.1 |
TC1100000950.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100008325.mm.1 |
NM_001271358.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008325.mm.1 |
NM_001271359.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008325.mm.1 |
NM_001271360.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008325.mm.1 |
NM_145429.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100008325.mm.1 |
TC1100000950.mm.1 |
MTA TC |