Assay Details
| Gene Symbol: Llgl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252532.1 | NP_001239461.1 | 3 - 4 | 330 | 89 | |
| NM_145438.2 | NP_663413.2 | 3 - 4 | 330 | 89 | ||
| GenBank mRNA | AK143486.1 | - | 3 - 4 | 317 | 89 | |
| AK143798.1 | - | 3 - 4 | 302 | 89 | ||
| AK167536.1 | - | 3 - 4 | 302 | 89 | ||
| AY033650.1 | - | 3 - 4 | 280 | 89 | ||
| BC037226.1 | - | 3 - 4 | 302 | 3584689 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Llgl2
Genomic Map