Assay Details
Gene Symbol: Stk24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145465.2 | NP_663440.1 | 7 - 8 | 1214 | 89 | |
GenBank mRNA | AK154959.1 | - | 7 - 8 | 1215 | 89 | |
AY188357.1 | - | 7 - 8 | 1224 | 89 | ||
BC004650.1 | - | 7 - 8 | 1046 | 3482980 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
