Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900006008.mm.1 |
NM_001081214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006008.mm.1 |
NM_001081214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006008.mm.1 |
TC1900000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006007.mm.1 |
NM_001081214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006007.mm.1 |
NM_001081214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006007.mm.1 |
TC1900000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006014.mm.1 |
NM_001081214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006014.mm.1 |
NM_001081214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006014.mm.1 |
TC1900000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006005.mm.1 |
NM_001081214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006005.mm.1 |
NM_001081214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006005.mm.1 |
TC1900000681.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900006011.mm.1 |
NM_001081214.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006011.mm.1 |
NM_001081214.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900006011.mm.1 |
TC1900000681.mm.1 |
MTA TC |