Assay Details
| Gene Symbol: Etfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145615.4 | NP_663590.3 | 1 - 2 | 211 | 103 | |
| GenBank mRNA | AK028118.1 | - | 1 - 2 | 212 | 103 | |
| AK032830.1 | - | 1 - 2 | 164 | 103 | ||
| AK133525.1 | - | 1 - 2 | 157 | 103 | ||
| AK167572.1 | - | 1 - 2 | 184 | 103 | ||
| BC003432.1 | - | 1 - 2 | 102 | 2646522 | 103 | |
| BC096645.1 | - | 1 - 2 | 70 | 1245213 | 103 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK028118.1,AK032830.1,AK133525.1,AK167572.1,BC003432.1,BC096645.1,NM_145615.4 transcripts.
Set Membership: |
|

Etfa
Genomic Map