Assay Details
| Gene Symbol: Etfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145615.4 | NP_663590.3 | 7 - 8 | 837 | 85 | |
| GenBank mRNA | AK028118.1 | - | 7 - 8 | 838 | 85 | |
| AK032830.1 | - | 7 - 8 | 790 | 85 | ||
| AK133525.1 | - | 7 - 8 | 783 | 85 | ||
| AK167572.1 | - | 7 - 8 | 810 | 85 | ||
| AK168321.1 | - | 7 - 8 | 798 | 85 | ||
| BC003432.1 | - | 7 - 8 | 728 | 2646522 | 85 | |
| BC096645.1 | - | 7 - 8 | 696 | 1245213 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Etfa
Genomic Map