Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900020429.mm.1 |
NM_001146046.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020429.mm.1 |
NM_001146046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020429.mm.1 |
NM_001146047.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020429.mm.1 |
NM_001146047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020429.mm.1 |
NM_145616.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020429.mm.1 |
NM_145616.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020429.mm.1 |
TC0900002435.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900020432.mm.1 |
NM_001146046.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020432.mm.1 |
NM_001146046.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020432.mm.1 |
NM_001146047.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020432.mm.1 |
NM_001146047.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020432.mm.1 |
NM_145616.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020432.mm.1 |
NM_145616.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900020432.mm.1 |
TC0900002435.mm.1 |
MTA TC |