Assay Details
| Gene Symbol: Alox12e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145684.1 | NP_663717.1 | 9 - 10 | 1313 | 97 | |
| GenBank mRNA | AK138743.1 | - | 9 - 10 | 1267 | 97 | |
| BC013751.1 | - | 9 - 10 | 1293 | 3596809 | 97 | |
| BC051047.1 | - | 9 - 10 | 1297 | 4457881 | 97 | |
| U39200.1 | - | 9 - 10 | 1254 | 97 | ||
| X99252.1 | - | 9 - 10 | 1313 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Alox12e
Genomic Map