Assay Details
Gene Symbol: Hist1h1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145713.4 | NP_663759.3 | 1 | 629 | 110 | |
GenBank mRNA | AK054269.1 | - | 627 | 110 | ||
AK155411.1 | - | 629 | 110 | |||
BC117045.1 | - | 1 | 626 | 40126364 | 110 | |
BC119173.1 | - | 1 | 626 | 8733922 | 110 |
Gene Symbol: E430020m11rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK088581.1 | - | 1 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28882160 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28882165 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28882161 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183508 | NM_145713.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2183508 | AK054269.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 583300 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87187 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183508 | NM_145713.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2183508 | AK054269.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P10683 | BC117045.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P10683 | BC119173.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P10683 | NM_145713.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P10683 | AK054269.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P501260 | AK155411.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P501260 | NM_145713.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P501260 | AK054269.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882176 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882165 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882177 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882169 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882180 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882168 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28882171 | - | - |
Whole Mouse Genome G4122A | A_51_P501260 | AK155411.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P501260 | NM_145713.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P501260 | AK054269.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P10683 | BC117045.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P10683 | BC119173.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P10683 | NM_145713.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P10683 | AK054269.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105903 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10816152 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28882160 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28882165 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28882161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882202 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882151 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882230 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28882203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10816138 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10816149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882197 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882181 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10786162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10816154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10816143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28882178 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10404035 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10404034 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10404033 | - | - |
Affymetrix Mouse Clariom S Array | PSR1300001407.mm.1 | NM_145713.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001407.mm.1 | TC1300000248.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001410.mm.1 | NM_145713.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001410.mm.1 | TC1300000248.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001409.mm.1 | NM_145713.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001409.mm.1 | TC1300000248.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001408.mm.1 | NM_145713.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001408.mm.1 | TC1300000248.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001411.mm.1 | NM_145713.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001411.mm.1 | TC1300000248.mm.1 | MTA TC |
Murine Genome U74v2 Set | 95388_at | - | - |
Murine Genome U74v2 Set | 95388_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 533275 | - | - |
ABI Mouse Genome Survey Microarray | 773027 | - | - |
ABI Mouse Genome Survey Microarray | 850696 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 3440446 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 104480017 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003440446 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1217657 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2475992 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1217657 | - | - |
mouse-6 v1.1 (Illumina) | scl11509.1.1_100-S | - | - |
mouse-6 v1.1 (Illumina) | scl0014957.2_1-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 104480017 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3440446 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0014957.2_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104480017 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3440446 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1217657 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1217657 | - | - |
