Assay Details
| Gene Symbol: Cenpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043344.1 | - | 5 - 6 | 682 | 66 | |
| AK145200.1 | - | 6 - 7 | 738 | 66 | ||
| AK161473.1 | - | 6 - 7 | 821 | 66 | ||
| AK166305.1 | - | 2 - 3 | 247 | 66 | ||
| BC030328.1 | - | 6 - 7 | 738 | 5376020 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpi
Genomic Map