Assay Details
| Gene Symbol: Mgat4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145926.3 | NP_666038.3 | 7 - 8 | 1101 | 59 | |
| GenBank mRNA | AB053218.1 | - | 7 - 8 | 803 | 59 | |
| AK155336.1 | - | 7 - 8 | 1102 | 59 | ||
| AK177701.1 | - | 2 - 3 | 115 | 59 | ||
| AK186986.1 | - | 2 - 3 | 115 | 59 | ||
| AK190141.1 | - | 115 | 59 | |||
| AK195601.1 | - | 2 - 3 | 115 | 59 | ||
| AK199018.1 | - | 2 - 3 | 115 | 59 | ||
| AK199371.1 | - | 114 | 59 | |||
| AK205336.1 | - | 2 - 3 | 115 | 59 | ||
| AK210581.1 | - | 2 - 3 | 115 | 59 | ||
| AK217144.1 | - | 2 - 3 | 115 | 59 | ||
| BC026638.1 | - | 2 - 3 | 209 | 4984907 | 59 | |
| BC031613.1 | - | 1 - 2 | 79 | 4216720 | 59 | |
| CX205743.1 | - | 4 - 5 | 361 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mgat4b
Genomic Map