Assay Details
| Gene Symbol: Mgat4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145926.3 | NP_666038.3 | 13 - 14 | 1818 | 63 | |
| GenBank mRNA | AB053218.1 | - | 13 - 14 | 1520 | 63 | |
| BC026638.1 | - | 8 - 9 | 926 | 4984907 | 63 | |
| BC031613.1 | - | 7 - 8 | 796 | 4216720 | 63 | |
| CX206051.1 | - | 5 - 6 | 577 | 63 | ||
| W66623.1 | - | 2 - 3 | 148 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mgat4b
Genomic Map