Assay Details
| Gene Symbol: AW549877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145930.2 | NP_666042.2 | 4 - 5 | 529 | 85 | |
| GenBank mRNA | AK045350.1 | - | 4 - 5 | 524 | 85 | |
| AK154940.1 | - | 4 - 5 | 524 | 85 | ||
| BC016504.1 | - | 3 - 4 | 407 | 3709876 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

AW549877
Genomic Map