Assay Details
Gene Symbol: Cacnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037099.2 | NP_001032176.1 | 7 - 8 | 888 | 65 | |
NM_001285426.1 | NP_001272355.1 | 7 - 8 | 672 | 65 | ||
NM_001285427.1 | NP_001272356.1 | 7 - 8 | 968 | 65 | ||
NM_001285428.1 | NP_001272357.1 | 6 - 7 | 597 | 65 | ||
NM_146123.3 | NP_666235.1 | 6 - 7 | 808 | 65 | ||
GenBank mRNA | AF068899.1 | - | 2 - 3 | 46 | 65 | |
AK032311.1 | - | 7 - 8 | 1112 | 65 | ||
AK038633.1 | - | 7 - 8 | 924 | 65 | ||
AK043850.1 | - | 6 - 7 | 751 | 65 | ||
AK079616.1 | - | 6 - 7 | 758 | 65 | ||
AK147338.1 | - | 7 - 8 | 862 | 65 | ||
AK158606.1 | - | 6 - 7 | 599 | 65 | ||
BC026479.1 | - | 6 - 7 | 594 | 4501980 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
