Assay Details
| Gene Symbol: Tpgs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148934.2 | NP_683736.1 | 1 - 2 | 378 | 99 | |
| GenBank mRNA | AF303106.1 | - | 1 - 2 | 378 | 99 | |
| AK153467.1 | - | 1 - 2 | 305 | 99 | ||
| BC138515.1 | - | 1 - 2 | 419 | 8861535 | 99 | |
| BC138516.1 | - | 1 - 2 | 419 | 8861536 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
multicellular organism development
spermatogenesis
sperm axoneme assembly
protein polyglutamylation
cell differentiation
adult behavior
vesicle localization
protein binding
microtubule binding
tubulin binding
tubulin-glutamic acid ligase activity
chemical synaptic transmission
multicellular organism development
spermatogenesis
sperm axoneme assembly
protein polyglutamylation
cell differentiation
adult behavior
vesicle localization
protein binding
microtubule binding
tubulin binding
tubulin-glutamic acid ligase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278163 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278196 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278160 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278198 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278161 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278193 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278157 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278162 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278194 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278197 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278195 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278158 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562593 | AF303106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562593 | AK153467.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562593 | BC138515.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562593 | BC138516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562593 | NM_148934.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562593 | AF303106.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562593 | AK153467.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562593 | BC138515.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562593 | BC138516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P562593 | NM_148934.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 942824 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96307 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103683 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P562593 | AF303106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P562593 | AK153467.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P562593 | BC138515.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P562593 | BC138516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P562593 | NM_148934.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P562593 | AF303106.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P562593 | AK153467.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P562593 | BC138515.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P562593 | BC138516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P562593 | NM_148934.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P562593 | AF303106.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P562593 | AK153467.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P562593 | BC138515.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P562593 | BC138516.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P562593 | NM_148934.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P562593 | AF303106.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P562593 | AK153467.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P562593 | BC138515.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P562593 | BC138516.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P562593 | NM_148934.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278161 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278171 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278169 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27278151 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103683 | - | - |
| Whole Mouse Genome G4122A | A_52_P562593 | AF303106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P562593 | AK153467.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P562593 | BC138515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P562593 | BC138516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P562593 | NM_148934.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P562593 | AF303106.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P562593 | AK153467.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P562593 | BC138515.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P562593 | BC138516.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P562593 | NM_148934.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P136406 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103683 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08589180 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08589163 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08589178 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278193 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278161 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278195 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278160 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278157 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278198 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278196 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278158 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278194 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278197 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278163 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27278162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27278261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08589156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08903410 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08937221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08300786 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278194 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08300781 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08589180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08878733 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08300778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27278166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08937217 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10364453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364452 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10364454 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451981_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10364451 | - | - |
| Mouse Genome 430 2.0 Array | 1451981_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1451981_at | - | - |
| Mouse Genome 430 2.0 Array | 1439646_at | - | - |
| Mouse Genome 430 2.0 Array | 1439646_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 110012_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 110012_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1000005843.mm.1 | NM_148934.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005843.mm.1 | NM_148934.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005843.mm.1 | TC1000000809.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005842.mm.1 | TC1000000809.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005844.mm.1 | NM_148934.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005844.mm.1 | NM_148934.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005844.mm.1 | TC1000000809.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1000005845.mm.1 | NM_148934.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005845.mm.1 | NM_148934.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1000005845.mm.1 | TC1000000809.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451981_PM_at | - | - |
| Mouse Expression Set 430 | 1451981_at | - | - |
| Murine Genome U74v2 Set | 160161_PM_at | - | - |
| Murine Genome U74v2 Set | 160161_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451981_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451981_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451981_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451981_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 740945 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6220053 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6220053 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006220053 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2592095 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2592095 | - | - |
| mouse-6 v1.1 (Illumina) | scl38711.3.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6220053 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl38711.3.1_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6220053 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2592095 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2592095 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl38711.3.1_9-S | - | - |

Tpgs1
Genomic Map