Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900022906.mm.1 |
NM_148941.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022906.mm.1 |
NM_148941.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022906.mm.1 |
TC0900002767.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022902.mm.1 |
NM_148941.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022902.mm.1 |
NM_148941.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022902.mm.1 |
TC0900002767.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022903.mm.1 |
NM_148941.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022903.mm.1 |
NM_148941.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022903.mm.1 |
TC0900002767.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900022907.mm.1 |
NM_148941.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022907.mm.1 |
NM_148941.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900022907.mm.1 |
TC0900002767.mm.1 |
MTA TC |