Assay Details
Gene Symbol: Ltc4s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008521.1 | NP_032547.1 | 3 - 4 | 270 | 62 | |
GenBank mRNA | BC028760.1 | - | 3 - 4 | 270 | 1348372 | 62 |
BC038518.1 | - | 3 - 4 | 280 | 62 | ||
U27195.1 | - | 3 - 4 | 270 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
