Assay Details
| Gene Symbol: Ldb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077398.2 | NP_001070866.1 | 5 - 6 | 874 | 71 | |
| NM_001286348.1 | NP_001273277.1 | 5 - 6 | 874 | 71 | ||
| NM_010698.4 | NP_034828.3 | 5 - 6 | 874 | 71 | ||
| GenBank mRNA | AK146860.1 | - | 5 - 6 | 808 | 71 | |
| AK147160.1 | - | 5 - 6 | 784 | 71 | ||
| BC079611.1 | - | 5 - 6 | 767 | 30360903 | 71 | |
| U89487.1 | - | 5 - 6 | 763 | 71 | ||
| U89489.1 | - | 5 - 6 | 763 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldb2
Genomic Map