Assay Details
| Gene Symbol: Mapt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038609.2 | NP_001033698.1 | 7 - 8 | 705 | 60 | |
| NM_001285454.1 | NP_001272383.1 | 5 - 6 | 531 | 60 | ||
| NM_001285455.1 | NP_001272384.1 | 4 - 5 | 465 | 60 | ||
| NM_001285456.1 | NP_001272385.1 | 5 - 6 | 531 | 60 | ||
| NM_010838.4 | NP_034968.3 | 5 - 6 | 531 | 60 | ||
| GenBank mRNA | AF483518.1 | - | 4 - 5 | 346 | 60 | |
| AF483519.1 | - | 4 - 5 | 346 | 60 | ||
| AK078167.1 | - | 5 - 6 | 504 | 60 | ||
| BC014748.1 | - | 5 - 6 | 469 | 4504850 | 60 | |
| CO432693.1 | - | 5 - 6 | 481 | 60 | ||
| M18775.1 | - | 5 - 6 | 532 | 60 | ||
| M18776.1 | - | 5 - 6 | 532 | 60 | ||
| U12914.1 | - | 7 - 8 | 589 | 60 | ||
| U12915.1 | - | 5 - 6 | 415 | 60 | ||
| U12916.1 | - | 4 - 5 | 349 | 60 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK078167.1 transcripts.
Set Membership: |
|

Mapt
Genomic Map