Assay Details
| Gene Symbol: Mbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013594.2 | NP_038622.2 | 9 - 10 | 1156 | 66 | |
| GenBank mRNA | AF072240.1 | - | 10 - 11 | 1135 | 66 | |
| AK004624.2 | - | 9 - 10 | 1157 | 66 | ||
| AK032392.1 | - | 1 - 2 | 100 | 66 | ||
| AK032535.1 | - | 6 - 7 | 654 | 66 | ||
| AK156401.1 | - | 10 - 11 | 1147 | 66 | ||
| AK166042.1 | - | 5 - 6 | 476 | 66 | ||
| BC069837.1 | - | 9 - 10 | 1172 | 30435760 | 66 | |
More Information
Set Membership: |
|

Mbd1
Genomic Map