Assay Details
| Gene Symbol: Bcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020486.2 | NP_065232.1 | 2 - 3 | 248 | 91 | |
| GenBank mRNA | AB035511.1 | - | 2 - 3 | 209 | 91 | |
| AF109160.1 | - | 2 - 3 | 248 | 91 | ||
| AF221507.1 | - | 2 - 3 | 198 | 91 | ||
| AF346663.1 | - | 2 - 3 | 186 | 91 | ||
| AK040564.1 | - | 2 - 3 | 232 | 91 | ||
| AK054001.1 | - | 2 - 3 | 229 | 91 | ||
| AK075704.1 | - | 2 - 3 | 213 | 91 | ||
| AK158439.1 | - | 2 - 3 | 208 | 91 | ||
| BC004826.1 | - | 2 - 3 | 232 | 2616148 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcam
Genomic Map