Assay Details
| Gene Symbol: Lias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024471.4 | NP_077791.1 | 4 - 5 | 523 | 92 | |
| GenBank mRNA | AB057731.1 | - | 4 - 5 | 422 | 92 | |
| AK031851.1 | - | 4 - 5 | 495 | 92 | ||
| AK045167.1 | - | 4 - 5 | 308 | 92 | ||
| AK048744.1 | - | 467 | 92 | |||
| AK049548.1 | - | 4 - 5 | 438 | 92 | ||
| AK077192.1 | - | 4 - 5 | 524 | 92 | ||
| BC002141.1 | - | 4 - 5 | 455 | 3484559 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lias
Genomic Map