Assay Details
| Gene Symbol: Lias | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024471.4 | NP_077791.1 | 5 - 6 | 682 | 66 | |
| GenBank mRNA | AB057731.1 | - | 5 - 6 | 581 | 66 | |
| AK031851.1 | - | 5 - 6 | 654 | 66 | ||
| AK045167.1 | - | 5 - 6 | 467 | 66 | ||
| AK048744.1 | - | 626 | 66 | |||
| AK049548.1 | - | 5 - 6 | 597 | 66 | ||
| AK077192.1 | - | 5 - 6 | 683 | 66 | ||
| BC002141.1 | - | 5 - 6 | 614 | 3484559 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lias
Genomic Map