Assay Details
| Gene Symbol: Cecr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144815.2 | NP_659064.1 | 4 - 5 | 546 | 95 | |
| GenBank mRNA | AK140387.1 | - | 4 - 5 | 567 | 95 | |
| BC014705.1 | - | 4 - 5 | 555 | 4238201 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cecr5
Genomic Map