Assay Details
| Gene Symbol: Ccdc92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144819.2 | NP_659068.1 | 2 - 3 | 356 | 91 | |
| GenBank mRNA | AK028192.1 | - | 2 - 3 | 333 | 91 | |
| AK136203.1 | - | 2 - 3 | 522 | 91 | ||
| AK165371.1 | - | 2 - 3 | 356 | 91 | ||
| BC021492.1 | - | 2 - 3 | 300 | 5345674 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc92
Genomic Map