Assay Details
| Gene Symbol: Senp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144851.5 | NP_659100.1 | 9 - 10 | 1313 | 88 | |
| GenBank mRNA | AK028565.1 | - | 9 - 10 | 1055 | 88 | |
| AK035581.1 | - | 9 - 10 | 1149 | 88 | ||
| AK054521.1 | - | 10 - 11 | 1445 | 88 | ||
| AK088597.1 | - | 9 - 10 | 1057 | 88 | ||
| BC023129.1 | - | 8 - 9 | 1027 | 3595534 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Senp1
Genomic Map