Assay Details
| Gene Symbol: Pja2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025309.1 | NP_001020480.1 | 6 - 7 | 1826 | 63 | |
| NM_144859.2 | NP_659108.1 | 5 - 6 | 1640 | 63 | ||
| GenBank mRNA | AF493070.1 | - | 6 - 7 | 1883 | 63 | |
| AK122282.1 | - | 6 - 7 | 1785 | 63 | ||
| AK144586.1 | - | 6 - 7 | 1813 | 63 | ||
| AK168371.1 | - | 5 - 6 | 1622 | 63 | ||
| BC004742.1 | - | 3 - 4 | 670 | 63 | ||
| BC017130.1 | - | 4 - 5 | 1566 | 4506259 | 63 | |
| BC119446.1 | - | 2 - 3 | 481 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pja2
Genomic Map