Assay Details
| Gene Symbol: Gatb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144896.4 | NP_659145.1 | 2 - 3 | 346 | 92 | |
| GenBank mRNA | AK028193.1 | - | 2 - 3 | 345 | 92 | |
| AK034702.1 | - | 2 - 3 | 347 | 92 | ||
| AK053978.1 | - | 2 - 3 | 349 | 92 | ||
| AK081800.1 | - | 2 - 3 | 350 | 92 | ||
| AK134831.1 | - | 2 - 3 | 281 | 92 | ||
| AK166231.1 | - | 2 - 3 | 357 | 92 | ||
| BC005709.1 | - | 2 - 3 | 348 | 3157973 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gatb
Genomic Map