Assay Details
| Gene Symbol: Gatb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144896.4 | NP_659145.1 | 6 - 7 | 887 | 102 | |
| GenBank mRNA | AK028193.1 | - | 6 - 7 | 886 | 102 | |
| AK034702.1 | - | 6 - 7 | 888 | 102 | ||
| AK053978.1 | - | 6 - 7 | 890 | 102 | ||
| AK081800.1 | - | 6 - 7 | 891 | 102 | ||
| AK134831.1 | - | 6 - 7 | 749 | 102 | ||
| BC005709.1 | - | 6 - 7 | 889 | 3157973 | 102 | |
More Information
Set Membership: |
|

Gatb
Genomic Map