Assay Details
| Gene Symbol: Gatb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144896.4 | NP_659145.1 | 8 - 9 | 1022 | 70 | |
| GenBank mRNA | AK028193.1 | - | 8 - 9 | 1021 | 70 | |
| AK034702.1 | - | 8 - 9 | 1023 | 70 | ||
| AK053978.1 | - | 8 - 9 | 1025 | 70 | ||
| AK081800.1 | - | 8 - 9 | 1026 | 70 | ||
| AK134831.1 | - | 8 - 9 | 884 | 70 | ||
| AK166231.1 | - | 8 - 9 | 1033 | 70 | ||
| AK182374.1 | - | 4 - 5 | 369 | 70 | ||
| BC005709.1 | - | 8 - 9 | 1024 | 3157973 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gatb
Genomic Map