Assay Details
Gene Symbol: Aldob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144903.2 | NP_659152.1 | 4 - 5 | 439 | 74 | |
GenBank mRNA | AK135385.1 | - | 4 - 5 | 468 | 74 | |
AK149395.1 | - | 4 - 5 | 456 | 74 | ||
AK149598.1 | - | 4 - 5 | 453 | 74 | ||
AK167334.1 | - | 4 - 5 | 456 | 74 | ||
AK167566.1 | - | 4 - 5 | 453 | 74 | ||
AK168600.1 | - | 4 - 5 | 453 | 74 | ||
AK168997.1 | - | 4 - 5 | 503 | 74 | ||
BC016435.1 | - | 4 - 5 | 435 | 4241025 | 74 | |
BC022113.1 | - | 4 - 5 | 412 | 5098524 | 74 | |
BC024056.1 | - | 4 - 5 | 445 | 5100067 | 74 | |
BC024112.1 | - | 4 - 5 | 444 | 5102118 | 74 | |
BC026577.1 | - | 4 - 5 | 404 | 4989521 | 74 | |
BC030724.1 | - | 4 - 5 | 441 | 5100784 | 74 | |
BC030725.1 | - | 4 - 5 | 445 | 5101686 | 74 | |
BC034169.1 | - | 4 - 5 | 445 | 4975382 | 74 | |
BC034171.1 | - | 4 - 5 | 422 | 4972521 | 74 | |
BC034172.1 | - | 4 - 5 | 409 | 4974489 | 74 | |
BC034173.1 | - | 4 - 5 | 421 | 4974553 | 74 | |
BC036130.1 | - | 4 - 5 | 422 | 4974071 | 74 | |
BC036131.1 | - | 4 - 5 | 439 | 4974877 | 74 | |
BC036132.1 | - | 4 - 5 | 439 | 4975336 | 74 | |
BC036133.1 | - | 4 - 5 | 451 | 4975573 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
