Assay Details
| Gene Symbol: Cnot6l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285511.1 | NP_001272440.1 | 5 - 6 | 665 | 67 | |
| NM_001285514.1 | NP_001272443.1 | 5 - 6 | 539 | 67 | ||
| NM_144910.2 | NP_659159.1 | 5 - 6 | 533 | 67 | ||
| NM_178854.4 | NP_849185.2 | 5 - 6 | 543 | 67 | ||
| GenBank mRNA | AK030112.1 | - | 5 - 6 | 541 | 67 | |
| AK151728.1 | - | 5 - 6 | 668 | 67 | ||
| AK207400.1 | - | 1 - 2 | 70 | 67 | ||
| BC018506.1 | - | 5 - 6 | 533 | 3598248 | 67 | |
| BC023923.1 | - | 1 - 2 | 55 | 67 | ||
| CF173938.1 | - | 5 - 6 | 515 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cnot6l
Genomic Map