Assay Details
| Gene Symbol: Daglb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144915.3 | NP_659164.2 | 13 - 14 | 1764 | 65 | |
| GenBank mRNA | AK081509.1 | - | 402 | 65 | ||
| AK086747.1 | - | 13 - 14 | 1676 | 65 | ||
| AK087884.1 | - | 13 - 14 | 1619 | 65 | ||
| AK136372.1 | - | 13 - 14 | 1622 | 65 | ||
| BC016105.1 | - | 13 - 14 | 1660 | 4921222 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Daglb
Genomic Map