Assay Details
| Gene Symbol: Hdac11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144919.2 | NP_659168.1 | 3 - 4 | 283 | 92 | |
| GenBank mRNA | AK045994.1 | - | 3 - 4 | 295 | 92 | |
| BC016208.1 | - | 3 - 4 | 283 | 4913322 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hdac11
Genomic Map