Assay Details
| Gene Symbol: Uroc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144940.2 | NP_659189.2 | 6 - 7 | 632 | 72 | |
| GenBank mRNA | AK149464.1 | - | 6 - 7 | 633 | 72 | |
| BC022133.1 | - | 6 - 7 | 615 | 5098655 | 72 | |
| BC033269.1 | - | 3 - 4 | 247 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Uroc1
Genomic Map