Assay Details
Gene Symbol: Alg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145939.2 | NP_666051.2 | 2 - 3 | 302 | 76 | |
GenBank mRNA | BC031893.1 | - | 2 - 3 | 297 | 5371807 | 76 |
CA538864.1 | - | 2 - 3 | 302 | 77 | ||
CX217502.1 | - | 2 - 3 | 249 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
