| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10412473 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412472 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412469 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412468 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412477 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412478 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412475 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412471 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412476 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412480 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412479 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412470 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10412474 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1433446_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1433444_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1441536_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1433443_PM_a_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1433445_PM_x_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10412466 |
- |
- |
| Mouse Genome 430 2.0 Array |
1433444_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1433446_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1433445_PM_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1441536_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1441536_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1433446_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1433443_PM_a_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1433444_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1433445_x_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1433443_a_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1300008504.mm.1 |
NM_145942.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300008504.mm.1 |
TC1300001353.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300008505.mm.1 |
NM_145942.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300008505.mm.1 |
TC1300001353.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300008503.mm.1 |
NM_145942.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300008503.mm.1 |
TC1300001353.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1433443_PM_a_at |
- |
- |
| Mouse Expression Set 430 |
1433445_PM_x_at |
- |
- |
| Mouse Expression Set 430 |
1433445_x_at |
- |
- |
| Mouse Expression Set 430 |
1433444_PM_at |
- |
- |
| Mouse Expression Set 430 |
1433444_at |
- |
- |
| Mouse Expression Set 430 |
1433446_at |
- |
- |
| Mouse Expression Set 430 |
1433446_PM_at |
- |
- |
| Mouse Expression Set 430 |
1433443_a_at |
- |
- |
| Mouse Expression Set 430 |
1441536_at |
- |
- |
| Mouse Expression Set 430 |
1441536_PM_at |
- |
- |
| Murine Genome U74v2 Set |
94325_at |
- |
- |
| Murine Genome U74v2 Set |
94325_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1433446_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1433444_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1433443_PM_a_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1433444_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1433445_x_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1433443_a_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1433445_PM_x_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1433446_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1441536_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1441536_PM_at |
- |
- |