Assay Details
| Gene Symbol: BC027231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145972.4 | NP_666084.1 | 6 - 7 | 983 | 88 | |
| GenBank mRNA | AK137274.1 | - | 5 - 6 | 1676 | 88 | |
| AK144612.1 | - | 6 - 7 | 964 | 88 | ||
| AK162291.1 | - | 5 - 6 | 825 | 88 | ||
| BC027231.1 | - | 6 - 7 | 986 | 3585175 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

BC027231
Genomic Map