Assay Details
| Gene Symbol: N4bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145974.3 | NP_666086.2 | 4 - 5 | 1517 | 58 | |
| GenBank mRNA | AK049249.1 | - | 4 - 5 | 1287 | 58 | |
| AK077848.1 | - | 4 - 5 | 1281 | 58 | ||
| AK141228.1 | - | 4 - 5 | 1311 | 58 | ||
| AK142500.1 | - | 4 - 5 | 1280 | 58 | ||
| AK170270.1 | - | 4 - 5 | 1315 | 58 | ||
| AK170521.1 | - | 4 - 5 | 1312 | 58 | ||
| BC005767.1 | - | 1 - 2 | 64 | 58 | ||
| BC006740.1 | - | 4 - 5 | 1230 | 3158173 | 58 | |
| BC055831.1 | - | 4 - 5 | 1517 | 4187601 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27795134 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208931 | BC055831.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208931 | BC005767.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208931 | AK170521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208931 | AK142500.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208931 | AK141228.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208931 | BC055831.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208931 | BC005767.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208931 | AK170521.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208931 | AK142500.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P208931 | AK141228.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625048 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546502 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74752 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372785 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123116 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116609 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208931 | BC055831.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208931 | BC005767.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208931 | AK170521.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208931 | AK142500.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208931 | AK141228.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208931 | BC055831.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208931 | BC005767.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208931 | AK170521.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208931 | AK142500.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P208931 | AK141228.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27795160 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116609 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02324271 | - | - |
| Whole Mouse Genome G4122A | A_51_P208931 | BC055831.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208931 | BC005767.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208931 | AK170521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208931 | AK142500.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208931 | AK141228.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208931 | BC055831.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208931 | BC005767.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208931 | AK170521.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208931 | AK142500.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P208931 | AK141228.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103391 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134477 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116609 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09539138 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09539128 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09539133 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09539137 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09567964 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09539132 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27795134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795125 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27795092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09539134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795135 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09554030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09539132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09554035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09133076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09236208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09556282 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09236203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795079 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795149 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09133072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09539126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09133069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09473315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09567964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09554018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09453820 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795096 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795089 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09150715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09539120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27795153 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10375887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385710 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385713 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385714 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385712 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385715 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10385711 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1451123_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10385709 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10375886 | - | - |
| Mouse Genome 430 2.0 Array | 1451123_at | - | - |
| Mouse Genome 430 2.0 Array | 1451123_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 212706_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 212706_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100024604.mm.1 | NM_145974.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024604.mm.1 | NM_145974.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100024604.mm.1 | TC1100002697.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1451123_at | - | - |
| Mouse Expression Set 430 | 1451123_PM_at | - | - |
| Murine Genome U74v2 Set | 165467_r_at | - | - |
| Murine Genome U74v2 Set | 167627_PM_f_at | - | - |
| Murine Genome U74v2 Set | 167627_f_at | - | - |
| Murine Genome U74v2 Set | 165467_PM_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451123_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1451123_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451123_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1451123_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3390458 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3390458 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003390458 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685223 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2685223 | - | - |
| mouse-6 v1.1 (Illumina) | scl40256.6.1_2-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3390458 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40256.6.1_2-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3390458 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2685222 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2685222 | - | - |

N4bp3
Genomic Map