Assay Details
| Gene Symbol: Arcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145985.4 | NP_666097.3 | 5 - 6 | 959 | 85 | |
| GenBank mRNA | AK028556.1 | - | 5 - 6 | 964 | 85 | |
| AK047995.1 | - | 5 - 6 | 998 | 85 | ||
| AK128973.1 | - | 5 - 6 | 964 | 85 | ||
| AK147747.1 | - | 5 - 6 | 994 | 85 | ||
| AK154066.1 | - | 963 | 85 | |||
| AK170005.1 | - | 1 - 2 | 146 | 85 | ||
| AK213335.1 | - | 1 - 2 | 55 | 85 | ||
| BC017124.1 | - | 5 - 6 | 954 | 4480874 | 85 | |
| BC023728.1 | - | 5 - 6 | 978 | 5351777 | 85 | |
| BC024127.1 | - | 4 - 5 | 579 | 85 | ||
| BC033387.1 | - | 5 - 6 | 959 | 5368060 | 85 | |
| BC034754.1 | - | 5 - 6 | 936 | 4954556 | 85 | |
| BC083152.1 | - | 952 | 30461105 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arcn1
Genomic Map